Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ANGEL2
Entrez Gene ID
90806
Gene Name
angel homolog 2
Gene Aliases
Ccr4d, KIAA0759L
Chromosome Location
Chr.1: 212992182 - 213015875 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001300753.1 NP_001287682.1 6 - 7 1392 102
NM_001300755.1 NP_001287684.1 6 - 7 1490 102
NM_001300757.1 NP_001287686.1 5 - 6 1145 102
NM_001300758.1 NP_001287687.1 5 - 6 1141 102
NM_144567.4 NP_653168.2 6 - 7 1467 102
XM_005273344.1 XP_005273401.1 6 - 7 1488 102
XM_005273345.1 XP_005273402.1 6 - 7 1534 102
XM_005273346.2 XP_005273403.1 6 - 7 1287 102
XM_005273347.3 XP_005273404.1 5 - 6 1159 102
XM_017002774.1 XP_016858263.1 6 - 7 1416 102
XM_017002775.1 XP_016858264.1 7 - 8 1715 102
XM_017002776.1 XP_016858265.1 7 - 8 1739 102
XM_017002777.1 XP_016858266.1 7 - 8 1785 102
XM_017002778.1 XP_016858267.1 7 - 8 1528 102
GenBank mRNA AF510741.1 - 5 - 6 1096 102
AK093947.1 - 1 - 2 1891 102
AK294479.1 - 6 - 7 1266 102
AK295116.1 - 6 - 7 1396 102
AL079275.1 - 6 - 7 1425 102
BC047469.1 - 5 - 6 1057 102

View Full Product Details