The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_144992.4 | NP_659429.4 | 19 - 20 | 2935 | 141 |
XM_005263897.2 | XP_005263954.1 | 19 - 20 | 2900 | 141 | |
XM_006712357.2 | XP_006712420.1 | 18 - 19 | 2649 | 141 | |
XM_006712359.3 | XP_006712422.1 | 10 - 11 | 1329 | 141 | |
XM_006712360.3 | XP_006712423.1 | 7 - 8 | 976 | 141 | |
XM_011510770.1 | XP_011509072.1 | 19 - 20 | 2900 | 141 | |
XM_011510771.2 | XP_011509073.1 | 18 - 19 | 2774 | 141 | |
XM_011510772.1 | XP_011509074.1 | 19 - 20 | 2900 | 141 | |
XM_017003560.1 | XP_016859049.1 | 13 - 14 | 1766 | 141 | |
XM_017003561.1 | XP_016859050.1 | 13 - 14 | 1772 | 141 | |
XM_017003562.1 | XP_016859051.1 | 13 - 14 | 1822 | 141 | |
XM_017003563.1 | XP_016859052.1 | 13 - 14 | 1772 | 141 | |
XM_017003564.1 | XP_016859053.1 | 19 - 20 | 2900 | 141 | |
GenBank mRNA | AK090786.1 | - | 6 - 7 | 771 | 141 |
AK093084.1 | - | 7 - 8 | 902 | 141 | |
AK295743.1 | - | 13 - 14 | 1830 | 141 | |
AL832761.1 | - | 19 - 20 | 2956 | 141 | |
AL834173.1 | - | 10 - 11 | 1334 | 141 | |
AL834385.1 | - | 9 - 10 | 1198 | 141 | |
BC150653.1 | - | 19 - 20 | 2875 | 141 |