The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_144992.4 | NP_659429.4 | 27 - 28 | 4004 | 109 |
XM_011510770.1 | XP_011509072.1 | 26 - 27 | 3918 | 109 | |
XM_011510772.1 | XP_011509074.1 | 26 - 27 | 3831 | 109 | |
XM_017003563.1 | XP_016859052.1 | 21 - 22 | 2841 | 109 |