Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- DNAJC27
- Entrez Gene ID
-
51277
- Gene Name
- DnaJ heat shock protein family (Hsp40) member C27
- Gene Aliases
-
RBJ, RabJS
- Chromosome Location
-
Chr.2: 24943636 - 24972094 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AL137731.1 |
- |
6 - 7 |
740 |
170 |
View Full Product Details