The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001098579.2 | NP_001092049.2 | 15 - 16 | 2121 | 83 |
NM_001100163.2 | NP_001093633.1 | 16 - 17 | 2399 | 83 | |
NM_001100167.2 | NP_001093637.1 | 16 - 17 | 2116 | 83 | |
NM_001206880.1 | NP_001193809.1 | 15 - 16 | 1834 | 83 | |
NM_001206881.1 | NP_001193810.1 | 14 - 15 | 2241 | 83 | |
NM_130385.3 | NP_569056.4 | 16 - 17 | 2452 | 83 | |
GenBank mRNA | AF081249.1 | - | 14 - 15 | 2137 | 83 |
AF081250.1 | - | 14 - 15 | 1865 | 83 | |
AK127209.1 | - | 16 - 17 | 2094 | 83 | |
AK296336.1 | - | 16 - 17 | 2216 | 83 | |
AK300358.1 | - | 15 - 16 | 2106 | 83 | |
AK304702.1 | - | 15 - 16 | 1468 | 83 | |
BC117127.1 | - | 16 - 17 | 1989 | 83 |