The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015295.2 | NP_056110.2 | 34 - 35 | 4541 | 155 |
XM_011525642.1 | XP_011523944.1 | 34 - 35 | 4541 | 155 | |
XM_011525643.2 | XP_011523945.1 | 34 - 35 | 4541 | 155 | |
XM_011525644.2 | XP_011523946.1 | 33 - 34 | 4449 | 155 | |
XM_017025684.1 | XP_016881173.1 | 35 - 36 | 4719 | 155 | |
GenBank mRNA | AB014550.1 | - | 8 - 9 | 883 | 155 |
AK126324.1 | - | 19 - 20 | 2172 | 155 | |
AL080138.1 | - | 5 - 6 | 521 | 155 | |
CR627458.1 | - | 23 - 24 | 2765 | 155 |