Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CAPN12
Entrez Gene ID
147968
Gene Name
calpain 12
Gene Aliases
-
Chromosome Location
Chr.19: 38730124 - 38744474 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_017026355.1 XP_016881844.1 13 - 14 2071 78
XM_017026356.1 XP_016881845.1 13 - 14 2071 78
XM_017026357.1 XP_016881846.1 14 - 15 1999 78
XM_017026359.1 XP_016881848.1 13 - 14 2071 78
XM_017026364.1 XP_016881853.1 13 - 14 1707 78

View Full Product Details