Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BSN
Entrez Gene ID
8927
Gene Name
bassoon presynaptic cytomatrix protein
Gene Aliases
ZNF231
Chromosome Location
Chr.3: 49554489 - 49671549 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003458.3 NP_003449.2 11 - 12 11994 103
GenBank mRNA AB007894.1 - 7 - 8 4816 103
AF052224.1 - 11 - 12 11995 103

View Full Product Details