The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014470.3 | NP_055285.1 | 3 - 4 | 424 | 74 |
GenBank mRNA | AB040147.1 | - | 3 - 4 | 402 | 74 |
AF498967.1 | - | 3 - 4 | 321 | 74 | |
AK292762.1 | - | 3 - 4 | 74 | ||
BC026356.1 | - | 3 - 4 | 424 | 74 | |
Y07923.1 | - | 3 - 4 | 74 |