The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001308267.1 | NP_001295196.1 | 13 - 14 | 2215 | 85 |
NM_001308268.1 | NP_001295197.1 | 14 - 15 | 2435 | 85 | |
NM_002847.4 | NP_002838.2 | 14 - 15 | 2329 | 85 | |
NM_130842.3 | NP_570857.2 | 13 - 14 | 2278 | 85 | |
NM_130843.3 | NP_570858.2 | 13 - 14 | 2242 | 85 | |
XM_011516446.1 | XP_011514748.1 | 14 - 15 | 2323 | 85 | |
XM_017012475.1 | XP_016867964.1 | 13 - 14 | 2334 | 85 | |
XM_017012476.1 | XP_016867965.1 | 14 - 15 | 2323 | 85 | |
GenBank mRNA | AB002385.2 | - | 14 - 15 | 2435 | 85 |
AF007555.1 | - | 14 - 15 | 2253 | 85 | |
BC034040.1 | - | 13 - 14 | 2213 | 85 | |
BC071570.1 | - | 14 - 15 | 2201 | 85 | |
U65065.1 | - | 13 - 14 | 2121 | 85 | |
U66702.1 | - | 14 - 15 | 2210 | 85 | |
U81561.1 | - | 13 - 14 | 2187 | 85 |