The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001308267.1 | NP_001295196.1 | 9 - 10 | 1662 | 116 |
NM_001308268.1 | NP_001295197.1 | 10 - 11 | 1882 | 116 | |
NM_002847.4 | NP_002838.2 | 10 - 11 | 1776 | 116 | |
NM_130842.3 | NP_570857.2 | 9 - 10 | 1725 | 116 | |
XM_011516446.1 | XP_011514748.1 | 10 - 11 | 1770 | 116 | |
XM_011516447.2 | XP_011514749.1 | 10 - 11 | 1770 | 116 | |
XM_011516448.2 | XP_011514750.1 | 10 - 11 | 1770 | 116 | |
XM_017012475.1 | XP_016867964.1 | 9 - 10 | 1781 | 116 | |
XM_017012476.1 | XP_016867965.1 | 10 - 11 | 1770 | 116 | |
GenBank mRNA | AB002385.2 | - | 10 - 11 | 1882 | 116 |
AF007555.1 | - | 10 - 11 | 1700 | 116 | |
BC071570.1 | - | 9 - 10 | 1648 | 116 | |
U66702.1 | - | 10 - 11 | 1657 | 116 | |
U81561.1 | - | 9 - 10 | 1634 | 116 |