The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014037.2 | NP_054756.2 | 2 - 3 | 647 | 76 |
XM_005258820.2 | XP_005258877.1 | 3 - 4 | 748 | 76 | |
XM_006723168.3 | XP_006723231.1 | 2 - 3 | 663 | 76 | |
XM_011526859.2 | XP_011525161.2 | 2 - 3 | 681 | 76 | |
XM_011526860.2 | XP_011525162.1 | 3 - 4 | 960 | 76 | |
XM_017026712.1 | XP_016882201.1 | 2 - 3 | 663 | 76 | |
GenBank mRNA | AF151977.1 | - | 1 - 2 | 452 | 76 |
AF265578.1 | - | 1 - 2 | 413 | 76 | |
AL136856.1 | - | 1 - 2 | 459 | 76 | |
BC034948.1 | - | 2 - 3 | 615 | 76 |