The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000334.4 | NP_000325.4 | 17 - 18 | 3394 | 72 |
GenBank mRNA | AY212253.1 | - | 17 - 18 | 3317 | 72 |
M81758.1 | - | 17 - 18 | 3394 | 72 |