The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007165.4 | NP_009096.2 | 3 - 4 | 322 | 66 |
GenBank mRNA | AK314815.1 | - | 3 - 4 | 315 | 66 |
BC004434.1 | - | 3 - 4 | 292 | 66 | |
BC009903.2 | - | 3 - 4 | 315 | 66 | |
BC015804.1 | - | 3 - 4 | 275 | 66 | |
BQ642367.1 | - | - | 395 | 66 |