The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007165.4 | NP_009096.2 | 8 - 9 | 740 | 127 |
GenBank mRNA | AK314815.1 | - | 8 - 9 | 733 | 127 |
BC004434.1 | - | 8 - 9 | 710 | 127 | |
BC009903.2 | - | 8 - 9 | 733 | 127 | |
BC015804.1 | - | 8 - 9 | 693 | 127 |