The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024867.3 | NP_079143.3 | 10 - 11 | 1632 | 83 |
XM_005248376.3 | XP_005248433.1 | 10 - 11 | 1737 | 83 | |
XM_005248377.3 | XP_005248434.1 | 10 - 11 | 1737 | 83 | |
XM_005248378.3 | XP_005248435.1 | 10 - 11 | 1726 | 83 | |
XM_011514135.2 | XP_011512437.1 | 10 - 11 | 1742 | 83 | |
XM_011514136.2 | XP_011512438.1 | 10 - 11 | 1741 | 83 | |
XM_011514137.2 | XP_011512439.1 | 10 - 11 | 1741 | 83 | |
XM_011514138.2 | XP_011512440.1 | 10 - 11 | 1742 | 83 | |
XM_011514139.2 | XP_011512441.1 | 10 - 11 | 1741 | 83 | |
XM_011514140.2 | XP_011512442.1 | 9 - 10 | 1476 | 83 | |
XM_011514141.2 | XP_011512443.1 | 11 - 12 | 1940 | 83 | |
XM_011514142.1 | XP_011512444.1 | 10 - 11 | 1737 | 83 | |
XM_017009880.1 | XP_016865369.1 | 9 - 10 | 1573 | 83 | |
XM_017009881.1 | XP_016865370.1 | 10 - 11 | 2596 | 83 | |
XM_017009882.1 | XP_016865371.1 | 10 - 11 | 1730 | 83 | |
GenBank mRNA | AL832607.1 | - | 3 - 4 | 540 | 83 |
EF553528.1 | - | 10 - 11 | 1582 | 83 |