The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024867.3 | NP_079143.3 | 2 - 3 | 271 | 129 |
NM_144722.3 | NP_653323.1 | 2 - 3 | 271 | 129 | |
XM_005248376.3 | XP_005248433.1 | 2 - 3 | 376 | 129 | |
XM_005248377.3 | XP_005248434.1 | 2 - 3 | 376 | 129 | |
XM_005248378.3 | XP_005248435.1 | 2 - 3 | 365 | 129 | |
XM_011514135.2 | XP_011512437.1 | 2 - 3 | 381 | 129 | |
XM_011514136.2 | XP_011512438.1 | 2 - 3 | 380 | 129 | |
XM_011514137.2 | XP_011512439.1 | 2 - 3 | 380 | 129 | |
XM_011514138.2 | XP_011512440.1 | 2 - 3 | 381 | 129 | |
XM_011514139.2 | XP_011512441.1 | 2 - 3 | 380 | 129 | |
XM_011514140.2 | XP_011512442.1 | 2 - 3 | 286 | 129 | |
XM_011514141.2 | XP_011512443.1 | 2 - 3 | 384 | 129 | |
XM_011514142.1 | XP_011512444.1 | 2 - 3 | 376 | 129 | |
XM_017009880.1 | XP_016865369.1 | 2 - 3 | 383 | 129 | |
XM_017009881.1 | XP_016865370.1 | 2 - 3 | 385 | 129 | |
XM_017009882.1 | XP_016865371.1 | 2 - 3 | 369 | 129 | |
GenBank mRNA | AK058124.1 | - | 2 - 3 | 196 | 129 |
BC110984.1 | - | 2 - 3 | 186 | 129 | |
EF553528.1 | - | 2 - 3 | 224 | 129 |