Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SPEF2
Entrez Gene ID
79925
Gene Name
sperm flagellar 2
Gene Aliases
CT122, KPL2
Chromosome Location
Chr.5: 35617773 - 35814611 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024867.3 NP_079143.3 6 - 7 904 107
NM_144722.3 NP_653323.1 6 - 7 904 107
XM_005248376.3 XP_005248433.1 6 - 7 1009 107
XM_005248377.3 XP_005248434.1 6 - 7 1009 107
XM_005248378.3 XP_005248435.1 6 - 7 998 107
XM_011514135.2 XP_011512437.1 6 - 7 1014 107
XM_011514136.2 XP_011512438.1 6 - 7 1013 107
XM_011514137.2 XP_011512439.1 6 - 7 1013 107
XM_011514138.2 XP_011512440.1 6 - 7 1014 107
XM_011514139.2 XP_011512441.1 6 - 7 1013 107
XM_011514140.2 XP_011512442.1 5 - 6 748 107
XM_011514141.2 XP_011512443.1 7 - 8 1212 107
XM_011514142.1 XP_011512444.1 6 - 7 1009 107
XM_017009880.1 XP_016865369.1 5 - 6 845 107
XM_017009881.1 XP_016865370.1 6 - 7 1868 107
XM_017009882.1 XP_016865371.1 6 - 7 1002 107
GenBank mRNA AK058124.1 - 6 - 7 829 107
BC110984.1 - 6 - 7 819 107
EF553528.1 - 6 - 7 855 107

View Full Product Details