Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HSPB7
Entrez Gene ID
27129
Gene Name
heat shock protein family B (small) member 7
Gene Aliases
cvHSP
Chromosome Location
Chr.1: 16014028 - 16018796 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_011541250.2 XP_011539552.1 1 - 2 1034 109
GenBank mRNA CR749419.1 - 1 - 2 281 109

View Full Product Details