The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015132.4 | NP_055947.1 | 18 - 19 | 2055 | 158 |
XM_005249672.1 | XP_005249729.1 | 18 - 19 | 2098 | 158 | |
XM_011515229.1 | XP_011513531.1 | 18 - 19 | 2098 | 158 | |
XM_011515230.2 | XP_011513532.1 | 19 - 20 | 2145 | 158 | |
XM_011515231.2 | XP_011513533.1 | 20 - 21 | 2204 | 158 | |
XM_011515233.2 | XP_011513535.1 | 19 - 20 | 2146 | 158 | |
XM_017011893.1 | XP_016867382.1 | 18 - 19 | 2065 | 158 | |
XM_017011894.1 | XP_016867383.1 | 19 - 20 | 2545 | 158 | |
XM_017011895.1 | XP_016867384.1 | 19 - 20 | 2117 | 158 | |
XM_017011896.1 | XP_016867385.1 | 19 - 20 | 4058 | 158 | |
XM_017011897.1 | XP_016867386.1 | 18 - 19 | 1986 | 158 | |
GenBank mRNA | AB018256.1 | - | 18 - 19 | 2048 | 158 |
AF121862.1 | - | 1 - 2 | 58 | 158 | |
AF420470.1 | - | 18 - 19 | 2023 | 158 | |
AK023740.1 | - | 13 - 14 | 1300 | 158 | |
AK026643.1 | - | 2 - 3 | 221 | 158 | |
AK294582.1 | - | 20 - 21 | 2010 | 158 | |
AK307453.1 | - | 1 - 2 | 187 | 158 | |
AK315135.1 | - | 18 - 19 | 2041 | 158 | |
BC045667.1 | - | 18 - 19 | 2025 | 158 |