The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015132.4 | NP_055947.1 | 2 - 3 | 334 | 149 |
XM_005249672.1 | XP_005249729.1 | 2 - 3 | 344 | 149 | |
XM_005249673.4 | XP_005249730.1 | 2 - 3 | 344 | 149 | |
XM_011515229.1 | XP_011513531.1 | 2 - 3 | 344 | 149 | |
XM_011515230.2 | XP_011513532.1 | 3 - 4 | 391 | 149 | |
XM_011515231.2 | XP_011513533.1 | 4 - 5 | 450 | 149 | |
XM_011515233.2 | XP_011513535.1 | 2 - 3 | 325 | 149 | |
XM_017011893.1 | XP_016867382.1 | 2 - 3 | 344 | 149 | |
XM_017011894.1 | XP_016867383.1 | 3 - 4 | 791 | 149 | |
XM_017011895.1 | XP_016867384.1 | 2 - 3 | 329 | 149 | |
XM_017011896.1 | XP_016867385.1 | 2 - 3 | 327 | 149 | |
XM_017011897.1 | XP_016867386.1 | 2 - 3 | 265 | 149 | |
GenBank mRNA | AB018256.1 | - | 2 - 3 | 294 | 149 |
AF420470.1 | - | 2 - 3 | 302 | 149 | |
AK001861.1 | - | 3 - 4 | 298 | 149 | |
AK315135.1 | - | 2 - 3 | 320 | 149 | |
AL353943.1 | - | 2 - 3 | 310 | 149 | |
BC045667.1 | - | 2 - 3 | 304 | 149 | |
BG718588.1 | - | 2 - 3 | 328 | 149 | |
BX503974.1 | - | 2 - 3 | 335 | 149 | |
DB050735.1 | - | 2 - 3 | 345 | 149 |