The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016616.4 | NP_057700.3 | 6 - 7 | 645 | 110 |
GenBank mRNA | AF202051.2 | - | 5 - 6 | 374 | 110 |
AF305596.1 | - | 5 - 6 | 343 | 110 | |
AK313599.1 | - | 5 - 6 | 385 | 110 | |
BC036816.1 | - | 6 - 7 | 645 | 110 | |
GQ472221.1 | - | 6 - 7 | 645 | 110 |