The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001278273.1 | NP_001265202.1 | 9 - 10 | 930 | 143 |
NM_182691.2 | NP_872633.1 | 7 - 8 | 983 | 143 | |
NM_182692.2 | NP_872634.1 | 8 - 9 | 906 | 143 | |
XM_005250549.3 | XP_005250606.2 | 9 - 10 | 1165 | 143 | |
XM_005250550.3 | XP_005250607.1 | 8 - 9 | 910 | 143 | |
XM_005250551.4 | XP_005250608.1 | 9 - 10 | 958 | 143 | |
XM_006716098.2 | XP_006716161.1 | 9 - 10 | 925 | 143 | |
XM_011516536.2 | XP_011514838.1 | 7 - 8 | 921 | 143 | |
XM_011516538.2 | XP_011514840.1 | 8 - 9 | 794 | 143 | |
XM_017012560.1 | XP_016868049.1 | 9 - 10 | 980 | 143 | |
XM_017012561.1 | XP_016868050.1 | 8 - 9 | 830 | 143 | |
XM_017012562.1 | XP_016868051.1 | 9 - 10 | 1164 | 143 | |
XM_017012563.1 | XP_016868052.1 | 9 - 10 | 1165 | 143 | |
XM_017012564.1 | XP_016868053.1 | 7 - 8 | 969 | 143 | |
XM_017012565.1 | XP_016868054.1 | 9 - 10 | 929 | 143 | |
XM_017012566.1 | XP_016868055.1 | 9 - 10 | 966 | 143 | |
GenBank mRNA | AY354201.1 | - | 8 - 9 | 971 | 143 |
BC035214.1 | - | 7 - 8 | 902 | 143 | |
BC068547.1 | - | 9 - 10 | 930 | 143 |