The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020762.2 | NP_065813.1 | 12 - 13 | 1590 | 104 |
XM_011538580.1 | XP_011536882.1 | 13 - 14 | 1698 | 104 | |
XM_011538581.2 | XP_011536883.1 | 13 - 14 | 3203 | 104 | |
GenBank mRNA | AK023899.1 | - | 2 - 3 | 320 | 104 |
BC053903.1 | - | 12 - 13 | 1590 | 104 |