The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024649.4 | NP_078925.3 | 1 - 2 | 60 | 73 |
GenBank mRNA | AF503941.1 | - | 1 - 2 | 60 | 73 |
AK095638.1 | - | 1 - 2 | 102 | 73 | |
AK294962.1 | - | 1 - 2 | 73 | 73 | |
AK296311.1 | - | 1 - 2 | 66 | 73 | |
AK302657.1 | - | 1 - 2 | 73 | 73 | |
BC047642.1 | - | 1 - 2 | 66 | 73 | |
BC109064.2 | - | 1 - 2 | 53 | 73 | |
BC109065.1 | - | 1 - 2 | 53 | 73 | |
BX647612.1 | - | 1 - 2 | 89 | 73 |