The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005700.4 | NP_005691.2 | 3 - 4 | 817 | 77 |
NM_130443.3 | NP_569710.2 | 3 - 4 | 820 | 77 | |
GenBank mRNA | AJ271216.1 | - | 3 - 4 | 416 | 77 |
AK021449.1 | - | 3 - 4 | 395 | 77 | |
AK222566.1 | - | 3 - 4 | 394 | 77 | |
AK222847.1 | - | 3 - 4 | 395 | 77 | |
AK304581.1 | - | 4 - 5 | 318 | 77 | |
AK315478.1 | - | 3 - 4 | 808 | 77 | |
AL833475.1 | - | 3 - 4 | 422 | 77 | |
BC001446.2 | - | 2 - 3 | 380 | 77 | |
BC007221.1 | - | 3 - 4 | 406 | 77 | |
BC014038.2 | - | 3 - 4 | 380 | 77 | |
BC024271.1 | - | 2 - 3 | 376 | 77 |