The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001134771.1 | NP_001128243.1 | 13 - 14 | 1827 | 90 |
NM_020708.4 | NP_065759.1 | 13 - 14 | 1845 | 90 | |
XM_017027981.1 | XP_016883470.1 | 13 - 14 | 1827 | 90 | |
GenBank mRNA | AB033002.1 | - | 13 - 14 | 1660 | 90 |
AF208159.1 | - | 13 - 14 | 1682 | 90 | |
AK289758.1 | - | 13 - 14 | 1804 | 90 | |
AK294059.1 | - | 9 - 10 | 1022 | 90 | |
AK295900.1 | - | 7 - 8 | 1041 | 90 | |
AK296331.1 | - | 7 - 8 | 743 | 90 | |
BC132668.1 | - | 13 - 14 | 1770 | 90 | |
BC132670.1 | - | 13 - 14 | 1770 | 90 |