The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014252.3 | NP_055067.1 | 4 - 5 | 773 | 130 |
GenBank mRNA | AF112968.1 | - | 4 - 5 | 610 | 130 |
AF177333.1 | - | 4 - 5 | 575 | 130 | |
AK296862.1 | - | 5 - 6 | 425 | 130 | |
AK313602.1 | - | 4 - 5 | 655 | 130 | |
BC002702.2 | - | 4 - 5 | 687 | 130 |