The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001301128.1 | NP_001288057.1 | 8 - 9 | 1481 | 75 |
NM_012301.3 | NP_036433.2 | 8 - 9 | 1481 | 75 | |
XM_011516718.1 | XP_011515020.1 | 8 - 9 | 1712 | 75 | |
XM_011516728.1 | XP_011515030.1 | 4 - 5 | 321 | 75 | |
XM_017012845.1 | XP_016868334.1 | 8 - 9 | 1712 | 75 | |
XM_017012846.1 | XP_016868335.1 | 8 - 9 | 1712 | 75 | |
XM_017012848.1 | XP_016868337.1 | 7 - 8 | 4997 | 75 | |
XM_017012849.1 | XP_016868338.1 | 7 - 8 | 4997 | 75 | |
GenBank mRNA | AB014605.2 | - | 8 - 9 | 1462 | 75 |
AF038563.1 | - | 8 - 9 | 1481 | 75 | |
AK297353.1 | - | 8 - 9 | 1466 | 75 | |
AK297580.1 | - | 7 - 8 | 1040 | 75 | |
AK298988.1 | - | 8 - 9 | 1356 | 75 | |
AK308685.1 | - | 4 - 5 | 302 | 75 | |
AK309012.1 | - | 4 - 5 | 323 | 75 | |
BC150277.1 | - | 8 - 9 | 1462 | 75 |