The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001127357.1 | NP_001120829.1 | 11 - 12 | 1493 | 143 |
NM_001127358.1 | NP_001120830.1 | 11 - 12 | 1570 | 143 | |
NM_020432.4 | NP_065165.3 | 10 - 11 | 1481 | 143 | |
XM_005250508.4 | XP_005250565.1 | 12 - 13 | 1545 | 143 | |
XM_005250509.4 | XP_005250566.1 | 12 - 13 | 1470 | 143 | |
XM_011516422.2 | XP_011514724.1 | 12 - 13 | 1545 | 143 | |
XM_011516423.2 | XP_011514725.1 | 11 - 12 | 1326 | 143 | |
XM_011516424.2 | XP_011514726.1 | 7 - 8 | 2008 | 143 | |
XM_017012458.1 | XP_016867947.1 | 11 - 12 | 1570 | 143 | |
XM_017012459.1 | XP_016867948.1 | 10 - 11 | 1351 | 143 | |
GenBank mRNA | AK090643.1 | - | 11 - 12 | 1549 | 143 |
AK225854.1 | - | 11 - 12 | 1557 | 143 | |
AK308289.1 | - | 11 - 12 | 1522 | 143 | |
AL050389.1 | - | 5 - 6 | 860 | 143 | |
AL136883.1 | - | 11 - 12 | 1461 | 143 | |
AL831969.2 | - | 12 - 13 | 1545 | 143 | |
BC127093.1 | - | 11 - 12 | 1409 | 143 | |
BC127094.1 | - | 10 - 11 | 1397 | 143 |