Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ERLIN1
Entrez Gene ID
10613
Gene Name
ER lipid raft associated 1
Gene Aliases
C10orf69, Erlin-1, KE04, KEO4, SPFH1, SPG62
Chromosome Location
Chr.10: 100150090 - 100186057 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001100626.1 NP_001094096.1 11 - 12 937 112
NM_006459.3 NP_006450.2 10 - 11 1181 112
XM_005269442.3 XP_005269499.1 11 - 12 967 112
XM_017015541.1 XP_016871030.1 11 - 12 951 112
GenBank mRNA AF064093.1 - 11 - 12 913 112
AK127017.1 - 4 - 5 3141 112
AK222480.1 - 11 - 12 909 112
AK298427.1 - 10 - 11 827 112
AK315580.1 - 11 - 12 909 112
BC031791.1 - 10 - 11 850 112

View Full Product Details