The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001102445.2 | NP_001095915.1 | 3 - 4 | 773 | 73 |
NM_001113380.1 | NP_001106851.1 | 2 - 3 | 348 | 73 | |
NM_001113381.1 | NP_001106852.1 | 2 - 3 | 384 | 73 | |
NM_005613.5 | NP_005604.1 | 2 - 3 | 664 | 73 | |
GenBank mRNA | AF493928.1 | - | 2 - 3 | 153 | 73 |
AK093959.1 | - | 2 - 3 | 277 | 73 | |
AK295540.1 | - | 3 - 4 | 508 | 73 | |
AK296240.1 | - | 2 - 3 | 192 | 73 | |
AK312250.1 | - | 2 - 3 | 276 | 73 | |
BC000737.1 | - | 2 - 3 | 238 | 73 | |
BC051869.1 | - | 2 - 3 | 250 | 73 | |
BT007025.1 | - | 2 - 3 | 153 | 73 | |
DA253378.1 | - | 3 - 4 | 508 | 73 | |
DA660018.1 | - | 2 - 3 | 433 | 73 | |
DA660695.1 | - | 2 - 3 | 420 | 73 | |
DQ346662.1 | - | 2 - 3 | 277 | 73 | |
DQ346663.1 | - | 2 - 3 | 492 | 73 | |
DQ346664.1 | - | 3 - 4 | 604 | 73 | |
EF054877.1 | - | 2 - 3 | 348 | 73 | |
U27768.1 | - | 2 - 3 | 250 | 73 |