Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RIMS1
Entrez Gene ID
22999
Gene Name
regulating synaptic membrane exocytosis 1
Gene Aliases
CORD7, RAB3IP2, RIM, RIM1
Chromosome Location
Chr.6: 71886703 - 72403143 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001168407.1 NP_001161879.1 10 - 11 1434 114
NM_001168408.1 NP_001161880.1 10 - 11 1434 114
NM_001168409.1 NP_001161881.1 10 - 11 1219 114
NM_001168410.1 NP_001161882.1 10 - 11 1506 114
NM_014989.5 NP_055804.2 15 - 16 3021 114
XM_005248688.3 XP_005248745.1 15 - 16 2957 114
XM_011535604.2 XP_011533906.2 15 - 16 2957 114
XM_011535611.2 XP_011533913.2 15 - 16 2957 114
XM_011535615.2 XP_011533917.2 15 - 16 2957 114
XM_017010516.1 XP_016866005.1 15 - 16 2957 114
XM_017010517.1 XP_016866006.1 15 - 16 2957 114
XM_017010518.1 XP_016866007.1 15 - 16 2957 114
XM_017010519.1 XP_016866008.1 15 - 16 2957 114
XM_017010520.1 XP_016866009.1 15 - 16 2957 114
XM_017010521.1 XP_016866010.1 15 - 16 2957 114
XM_017010522.1 XP_016866011.1 15 - 16 2957 114
XM_017010523.1 XP_016866012.1 15 - 16 2957 114
XM_017010524.1 XP_016866013.1 15 - 16 2957 114
XM_017010525.1 XP_016866014.1 15 - 16 2957 114
XM_017010526.1 XP_016866015.1 15 - 16 2957 114
XM_017010527.1 XP_016866016.1 15 - 16 2957 114
XM_017010528.1 XP_016866017.1 15 - 16 2957 114
XM_017010529.1 XP_016866018.1 15 - 16 2957 114
XM_017010530.1 XP_016866019.1 15 - 16 2957 114
XM_017010531.1 XP_016866020.1 15 - 16 2957 114
XM_017010532.1 XP_016866021.1 15 - 16 2957 114
XM_017010533.1 XP_016866022.1 15 - 16 2957 114
XM_017010534.1 XP_016866023.1 10 - 11 1530 114
XM_017010535.1 XP_016866024.1 10 - 11 1530 114
XM_017010536.1 XP_016866025.1 10 - 11 1527 114
XM_017010537.1 XP_016866026.1 10 - 11 1527 114
XM_017010538.1 XP_016866027.1 10 - 11 1635 114
XM_017010539.1 XP_016866028.1 10 - 11 1522 114
XM_017010540.1 XP_016866029.1 10 - 11 1521 114
XM_017010541.1 XP_016866030.1 10 - 11 1521 114
XM_017010542.1 XP_016866031.1 10 - 11 1521 114
XM_017010543.1 XP_016866032.1 10 - 11 1440 114
XM_017010544.1 XP_016866033.1 10 - 11 1523 114
XM_017010545.1 XP_016866034.1 10 - 11 1520 114
XM_017010546.1 XP_016866035.1 10 - 11 1633 114
XM_017010547.1 XP_016866036.1 10 - 11 1633 114
XM_017010548.1 XP_016866037.1 10 - 11 1518 114
XM_017010549.1 XP_016866038.1 10 - 11 1518 114
XM_017010550.1 XP_016866039.1 10 - 11 1901 114
XM_017010551.1 XP_016866040.1 10 - 11 1901 114
XM_017010552.1 XP_016866041.1 10 - 11 1758 114
XM_017010553.1 XP_016866042.1 10 - 11 1758 114
XM_017010554.1 XP_016866043.1 10 - 11 1754 114
XM_017010555.1 XP_016866044.1 10 - 11 1754 114
GenBank mRNA AB002338.1 - 16 - 17 2744 114
AB045726.1 - 16 - 17 2987 114
AB051866.1 - 16 - 17 2987 114
AF263305.1 - 3 - 4 375 114
AF263306.1 - 3 - 4 375 114
AF263307.1 - 3 - 4 375 114
AF263308.1 - 3 - 4 375 114
AF263309.1 - 3 - 4 375 114
AF263310.1 - 3 - 4 375 114
AK293828.1 - 10 - 11 1109 114
AK294868.1 - 10 - 11 1434 114
AK295001.1 - 10 - 11 1352 114
AK295177.1 - 10 - 11 1420 114
AK296303.1 - 10 - 11 1275 114
AK308658.1 - 10 - 11 1506 114
AK309185.1 - 10 - 11 1219 114
AK316108.1 - 10 - 11 1232 114
AY190519.1 - 15 - 16 2700 114
BC151853.1 - 16 - 17 2768 114
BC152435.1 - 16 - 17 2744 114

View Full Product Details