The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000051.3 | NP_000042.3 | 42 - 43 | 6578 | 78 |
XM_005271561.4 | XP_005271618.2 | 43 - 44 | 6639 | 78 | |
XM_005271562.4 | XP_005271619.2 | 42 - 43 | 6712 | 78 | |
XM_006718843.3 | XP_006718906.1 | 42 - 43 | 6345 | 78 | |
XM_006718845.1 | XP_006718908.1 | 17 - 18 | 2218 | 78 | |
XM_011542840.2 | XP_011541142.1 | 43 - 44 | 6433 | 78 | |
XM_011542842.2 | XP_011541144.1 | 41 - 42 | 6387 | 78 | |
XM_011542843.2 | XP_011541145.1 | 42 - 43 | 6926 | 78 | |
XM_011542844.2 | XP_011541146.1 | 42 - 43 | 6528 | 78 | |
XM_011542845.1 | XP_011541147.1 | 34 - 35 | 5041 | 78 | |
XM_017017789.1 | XP_016873278.1 | 43 - 44 | 7136 | 78 | |
XM_017017790.1 | XP_016873279.1 | 42 - 43 | 6533 | 78 | |
XM_017017791.1 | XP_016873280.1 | 42 - 43 | 6926 | 78 | |
GenBank mRNA | AK299843.1 | - | 4 - 5 | 409 | 78 |
BC137169.1 | - | 16 - 17 | 2216 | 78 | |
CR749436.1 | - | - | 1597 | 78 | |
U26455.1 | - | 16 - 17 | 2408 | 78 | |
U33841.1 | - | 43 - 44 | 6382 | 78 |