The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000051.3 | NP_000042.3 | 43 - 44 | 6736 | 157 |
XM_005271561.4 | XP_005271618.2 | 44 - 45 | 6797 | 157 | |
XM_005271562.4 | XP_005271619.2 | 43 - 44 | 6870 | 157 | |
XM_006718843.3 | XP_006718906.1 | 43 - 44 | 6503 | 157 | |
XM_006718845.1 | XP_006718908.1 | 18 - 19 | 2376 | 157 | |
XM_011542840.2 | XP_011541142.1 | 44 - 45 | 6591 | 157 | |
XM_011542842.2 | XP_011541144.1 | 42 - 43 | 6545 | 157 | |
XM_011542843.2 | XP_011541145.1 | 43 - 44 | 7084 | 157 | |
XM_011542844.2 | XP_011541146.1 | 43 - 44 | 6686 | 157 | |
XM_011542845.1 | XP_011541147.1 | 35 - 36 | 5199 | 157 | |
XM_017017789.1 | XP_016873278.1 | 44 - 45 | 7294 | 157 | |
XM_017017790.1 | XP_016873279.1 | 43 - 44 | 6691 | 157 | |
GenBank mRNA | AK299843.1 | - | 5 - 6 | 567 | 157 |
BC137169.1 | - | 17 - 18 | 2374 | 157 | |
CR749436.1 | - | - | 1755 | 157 | |
U26455.1 | - | 17 - 18 | 2566 | 157 | |
U33841.1 | - | 44 - 45 | 6540 | 157 |