The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000051.3 | NP_000042.3 | 17 - 18 | 3027 | 141 |
XM_005271561.4 | XP_005271618.2 | 18 - 19 | 3088 | 141 | |
XM_005271562.4 | XP_005271619.2 | 17 - 18 | 3161 | 141 | |
XM_006718843.3 | XP_006718906.1 | 17 - 18 | 2794 | 141 | |
XM_011542840.2 | XP_011541142.1 | 18 - 19 | 2882 | 141 | |
XM_011542842.2 | XP_011541144.1 | 16 - 17 | 2836 | 141 | |
XM_011542843.2 | XP_011541145.1 | 17 - 18 | 3375 | 141 | |
XM_011542844.2 | XP_011541146.1 | 17 - 18 | 2977 | 141 | |
XM_011542845.1 | XP_011541147.1 | 9 - 10 | 1490 | 141 | |
XM_017017789.1 | XP_016873278.1 | 18 - 19 | 3585 | 141 | |
XM_017017790.1 | XP_016873279.1 | 17 - 18 | 2982 | 141 | |
XM_017017791.1 | XP_016873280.1 | 17 - 18 | 3375 | 141 | |
XM_017017792.1 | XP_016873281.1 | 17 - 18 | 3375 | 141 | |
GenBank mRNA | U33841.1 | - | 18 - 19 | 2831 | 141 |
X91196.1 | - | 17 - 18 | 3375 | 141 |