Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NNMT
Entrez Gene ID
4837
Gene Name
nicotinamide N-methyltransferase
Gene Aliases
-
Chromosome Location
Chr.11: 114295813 - 114312516 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC000234.1 - 2 - 3 137 81

View Full Product Details