The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_005271621.4 | XP_005271678.1 | 2 - 3 | 4927 | 76 |
XM_006718892.3 | XP_006718955.1 | 2 - 3 | 457 | 76 | |
XM_011542926.2 | XP_011541228.1 | 2 - 3 | 454 | 76 | |
XM_017018034.1 | XP_016873523.1 | 3 - 4 | 525 | 76 | |
XM_017018035.1 | XP_016873524.1 | 3 - 4 | 522 | 76 | |
XM_017018036.1 | XP_016873525.1 | 2 - 3 | 992 | 76 | |
XM_017018037.1 | XP_016873526.1 | 3 - 4 | 1291 | 76 | |
XM_017018038.1 | XP_016873527.1 | 3 - 4 | 525 | 76 | |
XM_017018039.1 | XP_016873528.1 | 2 - 3 | 583 | 76 | |
XM_017018041.1 | XP_016873530.1 | 2 - 3 | 583 | 76 | |
XM_017018042.1 | XP_016873531.1 | 2 - 3 | 583 | 76 | |
XM_017018045.1 | XP_016873534.1 | 2 - 3 | 4924 | 76 |