Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TEX12
Entrez Gene ID
56158
Gene Name
testis expressed 12
Gene Aliases
-
Chromosome Location
Chr.11: 112167372 - 112172556 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031275.4 NP_112565.1 3 - 4 308 150
GenBank mRNA AF285600.1 - 2 - 3 214 150
BC029506.1 - 3 - 4 308 150
BI561514.1 - 3 - 4 312 150

View Full Product Details