Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CCDC84
Entrez Gene ID
338657
Gene Name
coiled-coil domain containing 84
Gene Aliases
DLNB14
Chromosome Location
Chr.11: 118998067 - 119015793 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_198489.2 NP_940891.1 9 - 10 860 117
NR_104049.1 9 - 10 888 117
NR_104051.1 9 - 10 1070 117
XM_011542799.1 XP_011541101.1 9 - 10 888 117
XM_011542800.2 XP_011541102.1 7 - 8 568 117
XM_017017652.1 XP_016873141.1 9 - 10 3642 117
GenBank mRNA AB094093.1 - 9 - 10 851 117
AW014772.1 - 2 - 3 178 117
BC126469.1 - 9 - 10 825 117
BC126471.1 - 9 - 10 825 117
BC144208.1 - 9 - 10 1035 117
BC144209.1 - 9 - 10 853 117
BU733365.1 - 2 - 3 117
DB565963.1 - 2 - 3 115 117

View Full Product Details