Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
USH2A
Entrez Gene ID
7399
Gene Name
usherin
Gene Aliases
RP39, US2, USH2, dJ1111A8.1
Chromosome Location
Chr.1: 215622894 - 216423396 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_206933.2 NP_996816.2 55 - 56 11325 62
GenBank mRNA AY481573.1 - 55 - 56 11325 62

View Full Product Details