The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001242338.2 | NP_001229267.2 | 11 - 12 | 1645 | 106 |
NM_017552.3 | NP_060022.2 | 11 - 12 | 1645 | 106 | |
NR_125717.1 | 11 - 12 | 1645 | 106 | ||
XM_005264372.3 | XP_005264429.2 | 12 - 13 | 1687 | 106 | |
XM_006712030.3 | XP_006712093.1 | 12 - 13 | 1687 | 106 | |
XM_006712031.3 | XP_006712094.1 | 12 - 13 | 1687 | 106 | |
XM_011532918.2 | XP_011531220.1 | 12 - 13 | 1687 | 106 | |
XM_011532919.2 | XP_011531221.1 | 12 - 13 | 1687 | 106 | |
XM_011532920.2 | XP_011531222.1 | 11 - 12 | 1645 | 106 | |
XM_011532921.2 | XP_011531223.1 | 12 - 13 | 1687 | 106 | |
XM_011532924.2 | XP_011531226.1 | 12 - 13 | 1687 | 106 | |
XM_011532925.2 | XP_011531227.1 | 12 - 13 | 1687 | 106 | |
XM_011532928.1 | XP_011531230.1 | 12 - 13 | 1687 | 106 | |
XM_011532929.2 | XP_011531231.1 | 12 - 13 | 1687 | 106 | |
XM_011532930.2 | XP_011531232.1 | 12 - 13 | 1687 | 106 | |
XM_011532931.2 | XP_011531233.1 | 12 - 13 | 1687 | 106 | |
XM_011532932.1 | XP_011531234.1 | 12 - 13 | 1687 | 106 | |
XM_017004375.1 | XP_016859864.1 | 11 - 12 | 1645 | 106 | |
GenBank mRNA | AK094821.1 | - | 11 - 12 | 1645 | 106 |
BC171846.1 | - | 11 - 12 | 1592 | 106 |