The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001040153.3 | NP_001035243.2 | 6 - 7 | 1244 | 60 |
NM_001242868.1 | NP_001229797.1 | 6 - 7 | 1753 | 60 | |
NM_001242869.1 | NP_001229798.1 | 5 - 6 | 1016 | 60 | |
NM_001242870.1 | NP_001229799.1 | 4 - 5 | 866 | 60 | |
NM_001242871.1 | NP_001229800.1 | 5 - 6 | 1292 | 60 | |
NM_144595.3 | NP_653196.1 | 5 - 6 | 1208 | 60 | |
XM_011534929.2 | XP_011533231.1 | 7 - 8 | 1633 | 60 | |
XM_011534930.1 | XP_011533232.1 | 6 - 7 | 1146 | 60 | |
GenBank mRNA | AK054608.1 | - | 5 - 6 | 988 | 60 |
AK094337.1 | - | 1 - 2 | 789 | 60 | |
AK289600.1 | - | 5 - 6 | 843 | 60 | |
AK289714.1 | - | 5 - 6 | 998 | 60 | |
AK294201.1 | - | 6 - 7 | 1244 | 60 | |
AK295423.1 | - | 5 - 6 | 1068 | 60 | |
AL834203.1 | - | 4 - 5 | 730 | 60 | |
AL834332.1 | - | 5 - 6 | 1001 | 60 | |
BC045177.1 | - | 5 - 6 | 1017 | 60 | |
BC111497.1 | - | 5 - 6 | 999 | 60 |