The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001201427.1 | NP_001188356.1 | 24 - 25 | 3119 | 86 |
NM_015345.3 | NP_056160.2 | 24 - 25 | 3180 | 86 | |
XM_006715039.3 | XP_006715102.3 | 25 - 26 | 3791 | 86 | |
XM_006715040.3 | XP_006715103.3 | 24 - 25 | 3763 | 86 | |
XM_006715042.2 | XP_006715105.1 | 25 - 26 | 5786 | 86 | |
XM_006715043.2 | XP_006715106.1 | 25 - 26 | 3163 | 86 | |
XM_006715045.3 | XP_006715108.1 | 25 - 26 | 4378 | 86 | |
XM_006715046.3 | XP_006715109.1 | 25 - 26 | 3556 | 86 | |
XM_017010630.1 | XP_016866119.1 | 25 - 26 | 4402 | 86 | |
GenBank mRNA | AB002379.2 | - | 24 - 25 | 3119 | 86 |
BX647839.1 | - | 8 - 9 | 3702 | 86 |