The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015295.2 | NP_056110.2 | 4 - 5 | 701 | 136 |
XM_011525642.1 | XP_011523944.1 | 4 - 5 | 701 | 136 | |
XM_011525643.2 | XP_011523945.1 | 4 - 5 | 701 | 136 | |
XM_011525644.2 | XP_011523946.1 | 3 - 4 | 609 | 136 | |
XM_017025684.1 | XP_016881173.1 | 5 - 6 | 879 | 136 | |
GenBank mRNA | BC006008.1 | - | 4 - 5 | 701 | 136 |