The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004724.3 | NP_004715.1 | 5 - 6 | 683 | 106 |
XM_017018558.1 | XP_016874047.1 | 5 - 6 | 623 | 106 | |
GenBank mRNA | AK300746.1 | - | 5 - 6 | 641 | 106 |
AK303871.1 | - | 4 - 5 | 362 | 106 | |
AK312925.1 | - | 5 - 6 | 661 | 106 | |
BC128264.1 | - | 5 - 6 | 611 | 106 | |
U54996.1 | - | 5 - 6 | 609 | 106 |