The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004724.3 | NP_004715.1 | 6 - 7 | 835 | 68 |
XM_017018559.1 | XP_016874048.1 | 6 - 7 | 791 | 68 | |
GenBank mRNA | AK303871.1 | - | 5 - 6 | 514 | 68 |
AK312925.1 | - | 6 - 7 | 813 | 68 | |
BC128264.1 | - | 6 - 7 | 763 | 68 | |
U54996.1 | - | 6 - 7 | 761 | 68 |