The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_198524.2 | NP_940926.1 | 2 - 3 | 149 | 113 |
XM_005254359.4 | XP_005254416.1 | 2 - 3 | 319 | 113 | |
XM_011521530.2 | XP_011519832.1 | 2 - 3 | 319 | 113 | |
XM_017022160.1 | XP_016877649.1 | 2 - 3 | 319 | 113 | |
XM_017022161.1 | XP_016877650.1 | 2 - 3 | 198 | 113 | |
XM_017022162.1 | XP_016877651.1 | 2 - 3 | 198 | 113 | |
GenBank mRNA | BC028119.1 | - | 2 - 3 | 127 | 113 |
HY164049.1 | - | 2 - 3 | 149 | 113 | |
KU178902.1 | - | 2 - 3 | 122 | 113 | |
KU178904.1 | - | 2 - 3 | 122 | 113 |