Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TTC5
Entrez Gene ID
91875
Gene Name
tetratricopeptide repeat domain 5
Gene Aliases
Strap
Chromosome Location
Chr.14: 20289131 - 20305994 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_138376.2 NP_612385.2 2 - 3 247 84
GenBank mRNA AK074553.1 - 2 - 3 230 84
BC008647.1 - 2 - 3 188 84
BC030822.1 - 2 - 3 191 84
BC053538.1 - 2 - 3 195 84
BX248066.1 - 2 - 3 184 84
BX640732.1 - 2 - 3 194 84
BX648584.1 - 2 - 3 202 84
DB051711.1 - 2 - 3 247 84

View Full Product Details