The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024529.4 | NP_078805.3 | 6 - 7 | 753 | 100 |
XM_006711537.3 | XP_006711600.1 | 6 - 7 | 677 | 100 | |
GenBank mRNA | AF312865.1 | - | 6 - 7 | 518 | 100 |
AK226038.1 | - | 6 - 7 | 650 | 100 | |
AK300929.1 | - | 6 - 7 | 612 | 100 | |
AK314772.1 | - | 6 - 7 | 511 | 100 | |
BC007325.2 | - | 6 - 7 | 536 | 100 | |
BC014351.1 | - | 2 - 3 | 144 | 100 | |
BC065037.1 | - | 6 - 7 | 590 | 100 |